Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ruvbl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019685.2 | NP_062659.1 | 3 - 4 | 464 | 108 | |
GenBank mRNA | AF100694.1 | - | 3 - 4 | 503 | 108 | |
AK132783.1 | - | 3 - 4 | 446 | 108 | ||
AK145062.1 | - | 413 | 108 | |||
AK146258.1 | - | 3 - 4 | 456 | 108 | ||
AK146374.1 | - | 3 - 4 | 457 | 108 | ||
AK156078.1 | - | 3 - 4 | 434 | 108 | ||
BC004718.1 | - | 3 - 4 | 430 | 3582426 | 108 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |