Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28047119 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28047200 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28047199 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28047197 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28047118 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28047117 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28047198 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28047121 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28047195 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28047194 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28047196 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28047193 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28047120 |
- |
- |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P230904 |
AB047850.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P230904 |
AF237687.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P230904 |
AK003622.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P230904 |
NM_019877.2 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P230904 |
AB040137.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P230904 |
AB047850.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P230904 |
AF237687.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P230904 |
AK003622.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P230904 |
NM_019877.2 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_51_P230904 |
AB040137.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2185397 |
AB047850.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2185397 |
AF237687.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2185397 |
AK003622.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2185397 |
AK187566.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2185397 |
NM_019877.2 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
596173 |
AK154596.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
596173 |
AK154959.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
596173 |
AY188357.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
596173 |
BC004650.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
596173 |
NM_145465.2 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
338226 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
624323 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
355822 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
795252 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
60360 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
175853 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P120968 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P159562 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P230904 |
AB047850.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P230904 |
AF237687.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P230904 |
AK003622.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P230904 |
NM_019877.2 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P230904 |
AB040137.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P230904 |
AB047850.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P230904 |
AF237687.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P230904 |
AK003622.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P230904 |
NM_019877.2 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_51_P230904 |
AB040137.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2185397 |
AB047850.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2185397 |
AF237687.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2185397 |
AK003622.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2185397 |
AK187566.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2185397 |
NM_019877.2 |
mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28047102 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28047111 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28047108 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28047101 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28047104 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28047128 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28047100 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28047126 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28047112 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28047130 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28047114 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28047105 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28047124 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28047134 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28047107 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P28047116 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P07454041 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P02403527 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P159562 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P120968 |
- |
- |
Whole Mouse Genome G4122A |
A_52_P180310 |
- |
- |
Whole Mouse Genome G4122A |
A_51_P230904 |
AB047850.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P230904 |
AF237687.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P230904 |
AK003622.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P230904 |
NM_019877.2 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P230904 |
AB040137.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P230904 |
AB047850.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P230904 |
AF237687.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P230904 |
AK003622.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P230904 |
NM_019877.2 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P230904 |
AB040137.1 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P113242 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P120968 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P159562 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P09453042 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P09453043 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P09315981 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P09211666 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P09315978 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P09211665 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P09315984 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28047119 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28047200 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28047121 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28047193 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28047199 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28047196 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28047117 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28047197 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28047198 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28047120 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28047195 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28047194 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P28047118 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28047207 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28047142 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28047119 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28047161 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28047084 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28047153 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28047198 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28047107 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28047191 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28047148 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28047184 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28047087 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28047193 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28047155 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28047210 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28047117 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28047111 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28047144 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28047146 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28047100 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28047101 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28047186 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28047140 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28047089 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28047157 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28047230 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28047169 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28047160 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28047189 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28047149 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28047218 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28047202 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28047211 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28047209 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28047201 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28047173 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28047188 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28047226 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28047172 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28047124 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28047126 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28047112 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28047195 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28047212 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28047187 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28047174 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28047167 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28047224 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28047083 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28047162 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28047085 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28047200 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28047116 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28047141 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28047151 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28047220 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28047136 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28047134 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28047181 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28047118 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28047139 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28047121 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28047216 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P28047096 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28047141 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28047111 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28047134 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28047226 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09650977 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28047113 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28047084 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28047157 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28047103 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28047132 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28047090 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09453043 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28047106 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09166094 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28047201 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09453042 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28047091 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09166091 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28047101 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09573891 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09701311 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28047114 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28047187 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28047144 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28047195 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09116159 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09573902 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28047100 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28047188 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09315978 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28047161 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09315975 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28047198 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09027999 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28047128 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28047089 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28047087 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28047228 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09087150 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28047178 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28047085 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09573892 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28047155 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28047121 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09237615 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28047083 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28047153 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28047219 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28047098 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09552614 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09211667 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09005554 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28047146 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28047221 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09612594 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28047160 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28047092 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28047189 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28047118 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28047184 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09298962 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28047095 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28047142 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09701309 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28047220 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28047097 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28047135 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28047148 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28047186 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09552622 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28047162 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09332427 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P28047130 |
- |
- |