Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0400017938.mm.1 |
NM_019969.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400017938.mm.1 |
NM_019969.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400017938.mm.1 |
TC0400002132.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400017939.mm.1 |
NM_019969.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400017939.mm.1 |
NM_019969.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400017939.mm.1 |
TC0400002132.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400017940.mm.1 |
NM_019969.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400017940.mm.1 |
NM_019969.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400017940.mm.1 |
TC0400002132.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400017942.mm.1 |
NM_019969.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400017942.mm.1 |
NM_019969.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400017942.mm.1 |
TC0400002132.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400017941.mm.1 |
NM_019969.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400017941.mm.1 |
NM_019969.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400017941.mm.1 |
TC0400002132.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400017943.mm.1 |
NM_019969.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400017943.mm.1 |
NM_019969.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400017943.mm.1 |
TC0400002132.mm.1 |
MTA TC |