Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DXBay18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025384.3 | NP_001020555.2 | 1 - 2 | 83 | 71 | |
GenBank mRNA | AK005439.1 | - | 1 - 2 | 84 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mus musculus Inventoried Probe spans exons |