Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myo15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103171.1 | NP_001096641.1 | 19 - 20 | 5451 | 73 | |
NM_010862.2 | NP_034992.2 | 19 - 20 | 5451 | 73 | ||
NM_182698.2 | NP_874357.2 | 18 - 19 | 1689 | 73 | ||
GenBank mRNA | AB014510.1 | - | 15 - 16 | 1454 | 73 | |
AF053130.1 | - | 17 - 18 | 1657 | 73 | ||
AF144095.1 | - | 19 - 20 | 5451 | 73 | ||
AY331132.1 | - | 19 - 20 | 5451 | 73 | ||
AY331133.1 | - | 18 - 19 | 1689 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mus musculus Probe spans exons |