Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700020392.mm.1 |
TC0700002254.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700020392.mm.1 |
NM_009493.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700020392.mm.1 |
NM_009493.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700020393.mm.1 |
TC0700002254.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700020393.mm.1 |
NM_009493.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700020393.mm.1 |
NM_009493.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700020266.mm.1 |
TC0700002233.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700020266.mm.1 |
NM_001105063.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700020266.mm.1 |
NM_001105063.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700020676.mm.1 |
TC0700002302.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700020676.mm.1 |
NM_001105156.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700020676.mm.1 |
NM_001105156.1 |
Refseq |