Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700020299.mm.1 |
TC0700002239.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700020299.mm.1 |
NM_001105066.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700020299.mm.1 |
NM_001105066.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700020442.mm.1 |
TC0700002262.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700020442.mm.1 |
NM_001105075.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700020442.mm.1 |
NM_001105075.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700020567.mm.1 |
TC0700002284.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700020567.mm.1 |
NM_009489.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700020567.mm.1 |
NM_009489.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700020300.mm.1 |
TC0700002239.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700020300.mm.1 |
NM_001105066.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700020300.mm.1 |
NM_001105066.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700020444.mm.1 |
TC0700002262.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700020444.mm.1 |
NM_001105075.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700020444.mm.1 |
NM_001105075.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700020565.mm.1 |
TC0700002284.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700020565.mm.1 |
NM_009489.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700020565.mm.1 |
NM_009489.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700020566.mm.1 |
TC0700002284.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700020566.mm.1 |
NM_009489.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700020566.mm.1 |
NM_009489.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700020443.mm.1 |
TC0700002262.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700020443.mm.1 |
NM_001105075.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700020443.mm.1 |
NM_001105075.1 |
Refseq |