Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1700006936.mm.1 |
NM_023124.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700006936.mm.1 |
NM_207648.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700006936.mm.1 |
NM_207648.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700006936.mm.1 |
TC1700002788.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700006934.mm.1 |
NM_023124.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700006934.mm.1 |
TC1700002788.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700006950.mm.1 |
NM_001198560.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700006950.mm.1 |
NM_001198560.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700006950.mm.1 |
NM_001198561.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700006950.mm.1 |
NM_001198561.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700006950.mm.1 |
NM_001201460.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700006950.mm.1 |
NM_010394.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700006950.mm.1 |
NM_010394.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700006950.mm.1 |
TC1700002789.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700006935.mm.1 |
NM_023124.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700006935.mm.1 |
NM_207648.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700006935.mm.1 |
NM_207648.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700006935.mm.1 |
TC1700002788.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700006947.mm.1 |
NM_001198560.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700006947.mm.1 |
NM_001198560.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700006947.mm.1 |
NM_001198561.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700006947.mm.1 |
NM_001198561.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700006947.mm.1 |
NM_001201460.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700006947.mm.1 |
NM_010394.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700006947.mm.1 |
NM_010394.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700006947.mm.1 |
TC1700002789.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700006949.mm.1 |
NM_001198560.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700006949.mm.1 |
NM_001198560.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700006949.mm.1 |
NM_001198561.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700006949.mm.1 |
NM_001198561.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700006949.mm.1 |
NM_001201460.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700006949.mm.1 |
NM_010394.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700006949.mm.1 |
NM_010394.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700006949.mm.1 |
TC1700002789.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700006937.mm.1 |
NM_023124.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700006937.mm.1 |
NM_207648.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700006937.mm.1 |
NM_207648.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700006937.mm.1 |
TC1700002788.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700006938.mm.1 |
NM_023124.5 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700006938.mm.1 |
NM_207648.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700006938.mm.1 |
NM_207648.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700006938.mm.1 |
TC1700002788.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700006948.mm.1 |
NM_001198560.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700006948.mm.1 |
NM_001198560.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700006948.mm.1 |
NM_001198561.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700006948.mm.1 |
NM_001198561.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700006948.mm.1 |
NM_001201460.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700006948.mm.1 |
NM_010394.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700006948.mm.1 |
NM_010394.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700006948.mm.1 |
TC1700002789.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1700006953.mm.1 |
NM_001198560.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700006953.mm.1 |
NM_001198560.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700006953.mm.1 |
NM_001201460.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700006953.mm.1 |
NM_010394.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700006953.mm.1 |
NM_010394.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1700006953.mm.1 |
TC1700002789.mm.1 |
MTA TC |