Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Gene 1.0 ST Array |
10570708 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10570674 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10570672 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10570713 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10570675 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10570652 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10570653 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10570708 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10570713 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10570672 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10570654 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10570652 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10570673 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10570675 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10570653 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10570673 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10570674 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10570694 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10570655 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10570654 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10570694 |
- |
- |
Affymetrix Mouse Gene 1.0 ST Array |
10570655 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10570671 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10570651 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10570671 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10570651 |
- |
- |
Mouse Genome 430 2.0 Array |
1436857_PM_at |
- |
- |
Mouse Genome 430 2.0 Array |
1436857_at |
- |
- |
Affymetrix Mouse Clariom S Array |
PSR0800001815.mm.1 |
NM_207658.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800001815.mm.1 |
NM_207658.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800001815.mm.1 |
TC0800000220.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800001785.mm.1 |
NM_183253.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800001785.mm.1 |
NM_183253.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800001785.mm.1 |
TC0800000211.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800001813.mm.1 |
NM_207658.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800001813.mm.1 |
NM_207658.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800001813.mm.1 |
TC0800000220.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800001786.mm.1 |
NM_183253.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800001786.mm.1 |
NM_183253.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800001786.mm.1 |
TC0800000211.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800001816.mm.1 |
NM_207658.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800001816.mm.1 |
NM_207658.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800001816.mm.1 |
TC0800000220.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800001814.mm.1 |
NM_207658.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800001814.mm.1 |
NM_207658.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800001814.mm.1 |
TC0800000220.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800001784.mm.1 |
NM_183253.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800001784.mm.1 |
NM_183253.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800001784.mm.1 |
TC0800000211.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0800001787.mm.1 |
NM_183253.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800001787.mm.1 |
NM_183253.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0800001787.mm.1 |
TC0800000211.mm.1 |
MTA TC |
Mouse Expression Set 430 |
1436857_PM_at |
- |
- |
Mouse Expression Set 430 |
1436857_PM_at |
- |
- |
Mouse Expression Set 430 |
1436857_at |
- |
- |
Mouse Expression Set 430 |
1436857_at |
- |
- |
Murine Genome U74v2 Set |
165492_at |
- |
- |
Murine Genome U74v2 Set |
165492_PM_at |
- |
- |
Murine Genome U74v2 Set |
165492_at |
- |
- |
Murine Genome U74v2 Set |
165492_PM_at |
- |
- |