Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Moap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022323.7 | NP_071718.1 | 1 - 2 | 165 | 111 | |
GenBank mRNA | BU614788.1 | - | 1 - 2 | 135 | 111 | |
BY344155.1 | - | 1 - 2 | 150 | 111 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Apoptosis Detect Genomic DNA Mus musculus Probe spans exons |