Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nat8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_023455.3 | NP_075944.1 | 1 - 2 | 103 | 78 | |
GenBank mRNA | AF163317.1 | - | 1 - 2 | 78 | ||
AK002784.1 | - | 1 - 2 | 45 | 78 | ||
BC019517.1 | - | 1 - 2 | 295 | 4236543 | 78 | |
BY081766.1 | - | 1 - 2 | 84 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |