Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010732.4 | NP_034862.1 | 1 - 2 | 322 | 104 | |
GenBank mRNA | AK036271.1 | - | 1 - 2 | 323 | 104 | |
AK039216.1 | - | 1 - 2 | 323 | 104 | ||
BC056458.1 | - | 1 - 2 | 64 | 5685881 | 104 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |