Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pydc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001162938.1 | NP_001156410.1 | 2 - 3 | 571 | 94 | |
GenBank mRNA | AK036671.1 | - | 2 - 3 | 341 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Mus musculus Probe spans exons |