Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Gene 1.1 ST Array |
10550770 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10560740 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10550770 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10560771 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10560730 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10560742 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10560787 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10560801 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10560797 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10560730 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10560789 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10560740 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10560789 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10550768 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10550768 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10560787 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10560783 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10560783 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10560797 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10550768 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10560789 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10560758 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10560742 |
- |
- |
Affymetrix Mouse Gene 1.1 ST Array |
10560742 |
- |
- |
Affymetrix Mouse Clariom S Array |
PSR0700022228.mm.1 |
TC0700002513.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700022228.mm.1 |
NM_001166723.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700022228.mm.1 |
NM_001166723.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700022228.mm.1 |
NM_001166749.1 |
Refseq |