Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Csrp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007792.4 | NP_031818.3 | 2 - 3 | 194 | 97 | |
GenBank mRNA | AF037208.1 | - | 2 - 3 | 194 | 97 | |
AK003328.1 | - | 2 - 3 | 193 | 97 | ||
AK010626.1 | - | 2 - 3 | 195 | 97 | ||
AK012647.1 | - | 2 - 3 | 192 | 97 | ||
BC012663.1 | - | 2 - 3 | 179 | 4210909 | 97 | |
BY333413.1 | - | 2 - 3 | 195 | 97 | ||
CK031156.1 | - | 2 - 3 | 180 | 97 | ||
D88792.1 | - | 1 - 2 | 120 | 97 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons |