Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pbx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001291508.1 | NP_001278437.1 | 2 - 3 | 932 | 91 | |
NM_001291509.1 | NP_001278438.1 | 2 - 3 | 932 | 91 | ||
NM_008783.3 | NP_032809.1 | 2 - 3 | 932 | 91 | ||
NM_183355.3 | NP_899198.1 | 2 - 3 | 932 | 91 | ||
GenBank mRNA | AB221563.1 | - | 2 - 3 | 267 | 91 | |
AF020196.1 | - | 2 - 3 | 623 | 91 | ||
AK084143.1 | - | 2 - 3 | 424 | 91 | ||
BC002244.1 | - | 2 - 3 | 158 | 3492658 | 91 | |
BC054376.1 | - | 2 - 3 | 893 | 91 | ||
BC058390.1 | - | 2 - 3 | 660 | 5701148 | 91 | |
HM369110.1 | - | 2 - 3 | 267 | 91 | ||
L27453.1 | - | 2 - 3 | 311 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons |