Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mxi1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008542.2 | NP_001008542.2 | 2 - 3 | 597 | 78 | |
NM_001008543.2 | NP_001008543.1 | 2 - 3 | 284 | 78 | ||
NM_010847.3 | NP_034977.1 | 2 - 3 | 437 | 78 | ||
GenBank mRNA | AK140465.1 | - | 2 - 3 | 387 | 78 | |
AK144219.1 | - | 2 - 3 | 479 | 78 | ||
AK154538.1 | - | 2 - 3 | 587 | 78 | ||
AK217176.1 | - | 1 - 2 | 46 | 78 | ||
BC064453.1 | - | 2 - 3 | 582 | 6810025 | 78 | |
D31824.1 | - | 2 - 3 | 437 | 78 | ||
L38821.1 | - | 2 - 3 | 290 | 78 | ||
L38822.1 | - | 2 - 3 | 309 | 78 | ||
U24673.1 | - | 2 - 3 | 270 | 78 | ||
U24674.1 | - | 2 - 3 | 219 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |