Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ankrd22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024204.6 | NP_077166.4 | 3 - 4 | 587 | 98 | |
GenBank mRNA | AK012869.1 | - | 3 - 4 | 600 | 98 | |
AK017360.1 | - | 3 - 4 | 588 | 98 | ||
AK017408.1 | - | 3 - 4 | 472 | 98 | ||
AK037242.1 | - | 3 - 4 | 470 | 98 | ||
BC003460.1 | - | 3 - 4 | 449 | 2648395 | 98 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |