Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uqcrc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025407.2 | NP_079683.2 | 6 - 7 | 782 | 74 | |
GenBank mRNA | AK010553.1 | - | 6 - 7 | 722 | 74 | |
AK013128.1 | - | 6 - 7 | 722 | 74 | ||
AK151764.1 | - | 6 - 7 | 724 | 74 | ||
AK160337.1 | - | 6 - 7 | 729 | 74 | ||
AK167911.1 | - | 6 - 7 | 722 | 74 | ||
AK168218.1 | - | 6 - 7 | 724 | 74 | ||
AK190382.1 | - | 1 - 2 | 91 | 74 | ||
AK214419.1 | - | 1 - 2 | 91 | 74 | ||
AK219902.1 | - | 2 - 3 | 169 | 74 | ||
BC030064.1 | - | 6 - 7 | 719 | 1400271 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |