Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plin3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025836.3 | NP_080112.1 | 1 - 2 | 59 | 104 | |
GenBank mRNA | AK004970.1 | - | 1 - 2 | 60 | 104 | |
AK012517.1 | - | 1 - 2 | 60 | 104 | ||
AK049537.1 | - | 1 - 2 | 60 | 104 | ||
AK050140.1 | - | 1 - 2 | 61 | 104 | ||
AK132313.1 | - | 60 | 104 | |||
AK167214.1 | - | 1 - 2 | 58 | 104 | ||
BC011116.1 | - | 1 - 2 | 55 | 4023354 | 104 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |