Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndufb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026612.3 | NP_080888.1 | 2 - 3 | 275 | 108 | |
GenBank mRNA | AK007495.1 | - | 2 - 3 | 251 | 108 | |
AK010642.1 | - | 2 - 3 | 282 | 108 | ||
AK019008.1 | - | 2 - 3 | 280 | 108 | ||
AK028334.1 | - | 284 | 108 | |||
AK216480.1 | - | 1 - 2 | 91 | 109 | ||
BC013510.1 | - | 2 - 3 | 285 | 4225177 | 108 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |