Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trmt12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026642.2 | NP_080918.2 | 1 | 1352 | 71 | |
GenBank mRNA | AK031429.1 | - | 1 | 1304 | 71 | |
AK076857.1 | - | 1 | 1322 | 71 | ||
AK079291.1 | - | 1 | 1310 | 71 | ||
BC052533.1 | - | 1 | 1322 | 5255779 | 71 | |
BC055850.1 | - | 1 | 1310 | 4193356 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Both primers and probe map within a single exon |