Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rasd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029182.1 | NP_083458.1 | 2 - 3 | 419 | 103 | |
GenBank mRNA | AK015898.1 | - | 2 - 3 | 650 | 103 | |
BC036988.1 | - | 2 - 3 | 430 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons |