Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029594.1 | NP_083870.1 | 2 - 3 | 219 | 130 | |
GenBank mRNA | AK005939.1 | - | 2 - 3 | 389 | 130 | |
AY509149.1 | - | 2 - 3 | 219 | 130 | ||
BC117082.1 | - | 2 - 3 | 218 | 40126401 | 130 | |
BC117084.1 | - | 2 - 3 | 218 | 40126403 | 130 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Mus musculus Probe spans exons |