Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sptssb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001286959.1 | NP_001273888.1 | 1 - 2 | 64 | 112 | |
NM_133675.3 | NP_598436.1 | 1 - 2 | 64 | 112 | ||
GenBank mRNA | AF365876.1 | - | 1 - 2 | 112 | ||
AK004017.1 | - | 1 - 2 | 66 | 112 | ||
AK010117.1 | - | 1 - 2 | 65 | 112 | ||
BC022628.1 | - | 1 - 2 | 44 | 4206004 | 112 | |
CX729774.1 | - | 1 - 2 | 38 | 112 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |