Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sptssb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164210.2 | NP_001157682.1 | 3 - 4 | 191 | 85 | |
NM_001286959.1 | NP_001273888.1 | 2 - 3 | 160 | 85 | ||
NM_133675.3 | NP_598436.1 | 2 - 3 | 160 | 85 | ||
GenBank mRNA | AF365876.1 | - | 2 - 3 | 162 | 85 | |
AK004017.1 | - | 2 - 3 | 162 | 85 | ||
AK009973.1 | - | 3 - 4 | 158 | 85 | ||
AK010117.1 | - | 2 - 3 | 161 | 85 | ||
BC022628.1 | - | 2 - 3 | 140 | 4206004 | 85 | |
CX729774.1 | - | 2 - 3 | 134 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |