Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Padi6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153106.2 | NP_694746.2 | 3 - 4 | 403 | 94 | |
GenBank mRNA | AF529423.1 | - | 3 - 4 | 376 | 94 | |
AK139616.1 | - | 3 - 4 | 404 | 94 | ||
BC053724.1 | - | 3 - 4 | 401 | 30051727 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |