Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pilra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153510.3 | NP_705730.1 | 2 - 3 | 637 | 103 | |
GenBank mRNA | AJ400844.1 | - | 535 | 103 | ||
AK041413.1 | - | 1 - 2 | 928 | 103 | ||
AK041670.1 | - | 2 - 3 | 549 | 103 | ||
AK170000.1 | - | 2 - 3 | 549 | 103 | ||
BC115565.2 | - | 522 | 40102015 | 103 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |