Array Name |
Probe IDs |
Targets |
Target Type |
Illumina mouse-6 v1.0 expression beadchip |
430204 |
AK005044.2 |
genbank_mrna_mouse |
Illumina mouse-6 v1.0 expression beadchip |
430204 |
AK042060.1 |
genbank_mrna_mouse |
Illumina mouse-6 v1.0 expression beadchip |
430204 |
AK043733.1 |
genbank_mrna_mouse |
Illumina mouse-6 v1.0 expression beadchip |
430204 |
AK047104.1 |
genbank_mrna_mouse |
Illumina mouse-6 v1.0 expression beadchip |
430204 |
AK049190.1 |
genbank_mrna_mouse |
Illumina mouse-6 v1.0 expression beadchip |
430204 |
AK080379.1 |
genbank_mrna_mouse |
Illumina mouse-6 v1.0 expression beadchip |
430204 |
AK088624.1 |
genbank_mrna_mouse |
Illumina mouse-6 v1.0 expression beadchip |
430204 |
AK150392.1 |
genbank_mrna_mouse |
Illumina mouse-6 v1.0 expression beadchip |
430204 |
AK151194.1 |
genbank_mrna_mouse |
Illumina mouse-6 v1.0 expression beadchip |
430204 |
AK152276.1 |
genbank_mrna_mouse |
Illumina mouse-6 v1.0 expression beadchip |
430204 |
AK152452.1 |
genbank_mrna_mouse |
Illumina mouse-6 v1.0 expression beadchip |
430204 |
AK152799.1 |
genbank_mrna_mouse |
Illumina mouse-6 v1.0 expression beadchip |
430204 |
AK153021.1 |
genbank_mrna_mouse |
Illumina mouse-6 v1.0 expression beadchip |
430204 |
AK153066.1 |
genbank_mrna_mouse |
Illumina mouse-6 v1.0 expression beadchip |
430204 |
AK154773.1 |
genbank_mrna_mouse |
Illumina mouse-6 v1.0 expression beadchip |
430204 |
AK172991.1 |
genbank_mrna_mouse |
Illumina mouse-6 v1.0 expression beadchip |
430204 |
AK197536.1 |
genbank_mrna_mouse |
Illumina mouse-6 v1.0 expression beadchip |
430204 |
BC024691.1 |
genbank_mrna_mouse |
Illumina mouse-6 v1.0 expression beadchip |
430204 |
NM_028791.5 |
refseq_mrna_mouse_NM |
Illumina mouseRef-8 v1.0 expression beadchip |
0000430204 |
- |
- |
mouse-6 v1.1 (Illumina) |
scl018775.4_0-S |
- |
- |
Illumina Sentrix mouse-6.v1 bead array |
430204 |
AK005044.2 |
genbank_mrna_mouse |
Illumina Sentrix mouse-6.v1 bead array |
430204 |
AK042060.1 |
genbank_mrna_mouse |
Illumina Sentrix mouse-6.v1 bead array |
430204 |
AK043733.1 |
genbank_mrna_mouse |
Illumina Sentrix mouse-6.v1 bead array |
430204 |
AK047104.1 |
genbank_mrna_mouse |
Illumina Sentrix mouse-6.v1 bead array |
430204 |
AK049190.1 |
genbank_mrna_mouse |
Illumina Sentrix mouse-6.v1 bead array |
430204 |
AK080379.1 |
genbank_mrna_mouse |
Illumina Sentrix mouse-6.v1 bead array |
430204 |
AK088624.1 |
genbank_mrna_mouse |
Illumina Sentrix mouse-6.v1 bead array |
430204 |
AK150392.1 |
genbank_mrna_mouse |
Illumina Sentrix mouse-6.v1 bead array |
430204 |
AK151194.1 |
genbank_mrna_mouse |
Illumina Sentrix mouse-6.v1 bead array |
430204 |
AK152276.1 |
genbank_mrna_mouse |
Illumina Sentrix mouse-6.v1 bead array |
430204 |
AK152452.1 |
genbank_mrna_mouse |
Illumina Sentrix mouse-6.v1 bead array |
430204 |
AK152799.1 |
genbank_mrna_mouse |
Illumina Sentrix mouse-6.v1 bead array |
430204 |
AK153021.1 |
genbank_mrna_mouse |
Illumina Sentrix mouse-6.v1 bead array |
430204 |
AK153066.1 |
genbank_mrna_mouse |
Illumina Sentrix mouse-6.v1 bead array |
430204 |
AK154773.1 |
genbank_mrna_mouse |
Illumina Sentrix mouse-6.v1 bead array |
430204 |
AK172991.1 |
genbank_mrna_mouse |
Illumina Sentrix mouse-6.v1 bead array |
430204 |
AK197536.1 |
genbank_mrna_mouse |
Illumina Sentrix mouse-6.v1 bead array |
430204 |
BC024691.1 |
genbank_mrna_mouse |
Illumina Sentrix mouse-6.v1 bead array |
430204 |
NM_028791.5 |
refseq_mrna_mouse_NM |
Illumina Sentrix MouseRef-8 Expression BeadChip |
scl018775.4_0-S |
- |
- |
Illumina Sentrix Mouse-6 Expression BeadChip (2) |
430204 |
AK005044.2 |
genbank_mrna_mouse |
Illumina Sentrix Mouse-6 Expression BeadChip (2) |
430204 |
AK042060.1 |
genbank_mrna_mouse |
Illumina Sentrix Mouse-6 Expression BeadChip (2) |
430204 |
AK043733.1 |
genbank_mrna_mouse |
Illumina Sentrix Mouse-6 Expression BeadChip (2) |
430204 |
AK047104.1 |
genbank_mrna_mouse |
Illumina Sentrix Mouse-6 Expression BeadChip (2) |
430204 |
AK049190.1 |
genbank_mrna_mouse |
Illumina Sentrix Mouse-6 Expression BeadChip (2) |
430204 |
AK080379.1 |
genbank_mrna_mouse |
Illumina Sentrix Mouse-6 Expression BeadChip (2) |
430204 |
AK088624.1 |
genbank_mrna_mouse |
Illumina Sentrix Mouse-6 Expression BeadChip (2) |
430204 |
AK150392.1 |
genbank_mrna_mouse |
Illumina Sentrix Mouse-6 Expression BeadChip (2) |
430204 |
AK151194.1 |
genbank_mrna_mouse |
Illumina Sentrix Mouse-6 Expression BeadChip (2) |
430204 |
AK152276.1 |
genbank_mrna_mouse |
Illumina Sentrix Mouse-6 Expression BeadChip (2) |
430204 |
AK152452.1 |
genbank_mrna_mouse |
Illumina Sentrix Mouse-6 Expression BeadChip (2) |
430204 |
AK152799.1 |
genbank_mrna_mouse |
Illumina Sentrix Mouse-6 Expression BeadChip (2) |
430204 |
AK153021.1 |
genbank_mrna_mouse |
Illumina Sentrix Mouse-6 Expression BeadChip (2) |
430204 |
AK153066.1 |
genbank_mrna_mouse |
Illumina Sentrix Mouse-6 Expression BeadChip (2) |
430204 |
AK154773.1 |
genbank_mrna_mouse |
Illumina Sentrix Mouse-6 Expression BeadChip (2) |
430204 |
AK172991.1 |
genbank_mrna_mouse |
Illumina Sentrix Mouse-6 Expression BeadChip (2) |
430204 |
AK197536.1 |
genbank_mrna_mouse |
Illumina Sentrix Mouse-6 Expression BeadChip (2) |
430204 |
BC024691.1 |
genbank_mrna_mouse |
Illumina Sentrix Mouse-6 Expression BeadChip (2) |
430204 |
NM_028791.5 |
refseq_mrna_mouse_NM |
MouseRef-8 v2 Expression BeadChip |
ILMN_2964485 |
- |
- |
Illumina MouseWG-6 v2.0 expression beadchip |
ILMN_2592926 |
- |
- |
Illumina MouseWG-6 v2.0 expression beadchip |
ILMN_2964485 |
- |
- |