Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gjc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175452.4 | NP_780661.2 | 1 - 2 | 84 | 95 | |
GenBank mRNA | AK046421.1 | - | 1 - 2 | 85 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mus musculus Inventoried Probe spans exons |