Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Syt15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_176931.2 | NP_795905.2 | 2 - 3 | 330 | 99 | |
NM_181529.4 | NP_852682.1 | 2 - 3 | 330 | 99 | ||
GenBank mRNA | AB109019.1 | - | 1 - 2 | 202 | 99 | |
AB109020.1 | - | 1 - 2 | 202 | 99 | ||
AK042641.1 | - | 2 - 3 | 331 | 99 | ||
AK054180.1 | - | 2 - 3 | 330 | 99 | ||
BB639494.1 | - | 2 - 3 | 331 | 99 | ||
BC042700.1 | - | 2 - 3 | 781 | 99 | ||
BC139313.1 | - | 2 - 3 | 241 | 8862335 | 99 | |
BC139314.1 | - | 2 - 3 | 242 | 8862336 | 99 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |