Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc35e2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177186.4 | NP_796160.1 | 1 - 2 | 318 | 95 | |
GenBank mRNA | AK080215.1 | - | 1 - 2 | 337 | 95 | |
BC058728.1 | - | 1 - 2 | 318 | 6407259 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Transporter Mammalian Gene Collection Mus musculus 1700 Probe spans exons |