Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bpi | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177850.3 | NP_808518.1 | 11 - 12 | 1287 | 106 | |
GenBank mRNA | AK033770.1 | - | 11 - 12 | 1289 | 106 | |
AY363993.1 | - | 11 - 12 | 1276 | 106 | ||
AY648037.1 | - | 11 - 12 | 1278 | 106 | ||
AY853179.1 | - | 11 - 12 | 1263 | 106 | ||
BC118049.1 | - | 11 - 12 | 1272 | 40106697 | 106 | |
BC118504.1 | - | 11 - 12 | 1272 | 40106694 | 106 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |