Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zbed4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_181412.3 | NP_852077.1 | 2 - 3 | 262 | 80 | |
GenBank mRNA | AK122335.1 | - | 2 - 3 | 232 | 80 | |
BC052174.1 | - | 1 - 2 | 165 | 6518433 | 80 | |
CD350815.1 | - | 2 - 3 | 249 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |