Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1300002209.mm.1 |
NM_011456.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300002209.mm.1 |
NM_011456.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300002209.mm.1 |
TC1300000368.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300002216.mm.1 |
NM_183197.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300002216.mm.1 |
NM_183197.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300002216.mm.1 |
TC1300000369.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300002208.mm.1 |
NM_011456.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300002208.mm.1 |
NM_011456.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300002208.mm.1 |
TC1300000368.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300002207.mm.1 |
NM_011456.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300002207.mm.1 |
NM_011456.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300002207.mm.1 |
TC1300000368.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300002218.mm.1 |
NM_183197.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300002218.mm.1 |
NM_183197.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300002218.mm.1 |
TC1300000369.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1300002217.mm.1 |
NM_183197.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300002217.mm.1 |
NM_183197.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1300002217.mm.1 |
TC1300000369.mm.1 |
MTA TC |