Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ms4a13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_198224.3 | NP_937867.2 | 5 - 6 | 615 | 112 | |
GenBank mRNA | AF544402.1 | - | 5 - 6 | 620 | 112 | |
AK006841.1 | - | 5 - 6 | 631 | 112 | ||
BC089483.1 | - | 5 - 6 | 615 | 6772390 | 112 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons |