Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Romo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025946.6 | NP_080222.1 | 1 | 254 | 110 | |
GenBank mRNA | AY028425.1 | - | 1 | 149 | 110 | |
EL963857.1 | - | 1 | 264 | 110 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Mus musculus Both primers and probe map within a single exon |