Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1400008506.mm.1 |
NM_009429.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400008506.mm.1 |
NM_009429.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400008506.mm.1 |
TC1400001061.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400008507.mm.1 |
NM_009429.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400008507.mm.1 |
NM_009429.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400008507.mm.1 |
TC1400001061.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400008511.mm.1 |
NM_009429.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400008511.mm.1 |
NM_009429.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400008511.mm.1 |
TC1400001061.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1400008512.mm.1 |
NM_009429.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400008512.mm.1 |
NM_009429.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1400008512.mm.1 |
TC1400001061.mm.1 |
MTA TC |