Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tgoln1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009443.3 | NP_033469.1 | 1 | 152 | 82 | |
GenBank mRNA | AK076586.1 | - | 1 | 153 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Both primers and probe map within a single exon |