Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Xlr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011725.4 | NP_035855.1 | 1 | 64 | 108 | |
GenBank mRNA | AK012549.1 | - | 1 | 55 | 108 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Amplicon spans exons and probe does not span exons |