Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcdhga6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_033589.1 | NP_291067.1 | 1 | 1353 | 103 | |
GenBank mRNA | AY013800.1 | - | 1 | 1353 | 103 | |
BC059927.1 | - | 1 | 1488 | 103 | ||
BC080852.1 | - | 1 | 1398 | 6844625 | 103 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Both primers and probe map within a single exon |