Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Glcci1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133236.3 | NP_573499.2 | 1 | 548 | 118 | |
GenBank mRNA | AY098636.1 | - | 1 | 594 | 115 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Mus musculus Inventoried Both primers and probe map within a single exon |