Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027046.3 | NP_081322.1 | 1 | 151 | 81 | |
GenBank mRNA | AK005741.1 | - | 1 | 87 | 81 | |
BC049741.1 | - | 1 | 151 | 6771599 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Both primers and probe map within a single exon |