Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Defa-rs4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005418.1 | NP_001005418.1 | 52 | 80 | ||
GenBank mRNA | AJ564861.1 | - | 52 | 80 |
Gene Symbol: Defa-rs2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AJ564864.1 | - | 52 | 80 | ||
AJ564865.1 | - | 52 | 80 | |||
AJ564866.1 | - | 52 | 80 | |||
AJ564867.1 | - | 52 | 80 |
Gene Symbol: Defa-rs7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AJ564872.1 | - | 52 | 80 | ||
AJ564873.1 | - | 52 | 80 | |||
AJ564875.1 | - | 52 | 80 | |||
AJ564876.1 | - | 52 | 80 | |||
AJ564877.1 | - | 52 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon |