Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Defa-rs10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007845.3 | NP_031871.1 | 172 | 70 | ||
GenBank mRNA | BC119615.1 | - | 172 | 40061719 | 70 | |
BC119616.1 | - | 172 | 40061721 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus Amplicon spans exons and probe does not span exons |