Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Jmjd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205068.1 | NP_001191997.1 | 2 - 3 | 637 | 76 | |
NM_178659.6 | NP_848774.1 | 2 - 3 | 637 | 76 | ||
NR_037992.1 | 2 - 3 | 637 | 76 | |||
GenBank mRNA | AK032477.1 | - | 2 - 3 | 611 | 76 | |
AK037954.1 | - | 2 - 3 | 636 | 76 | ||
AK038650.1 | - | 1 - 2 | 180 | 76 | ||
AK039116.1 | - | 2 - 3 | 315 | 76 | ||
AK047586.1 | - | 2 - 3 | 638 | 76 | ||
AK052144.1 | - | 2 - 3 | 623 | 76 | ||
AK079104.1 | - | 2 - 3 | 595 | 76 | ||
AK084669.1 | - | 2 - 3 | 595 | 76 | ||
AK145394.1 | - | 2 - 3 | 615 | 76 | ||
AK171919.1 | - | 2 - 3 | 592 | 76 | ||
BC049148.1 | - | 2 - 3 | 622 | 76 | ||
BC116707.1 | - | 2 - 3 | 568 | 40096667 | 76 | |
BC118020.1 | - | 2 - 3 | 567 | 76 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |