Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krt74 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_033444.1 | 6 - 7 | 1038 | 80 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | keratin 71 | Krt71 |
AA589543, Ca, Cal4, Cu, Krt2-6g, mK6irs, mK6irs1 |
-13.5kb | Sense | Chr. 15 - 101733949 - 101743097 on NCBI Build GRCm38 |
Downstream | keratin 72 | Krt72 |
AI507495, K6irs2, Krt2-35, Krt72-ps |
17.6kb | Sense | Chr. 15 - 101776560 - 101786458 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length between 71 and 85 Mus musculus Probe spans exons |