Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 5730416F02Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_033596.1 | 1 | 251 | 94 | ||
GenBank mRNA | AK017567.1 | - | 1 | 261 | 94 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
---|---|---|---|---|---|---|---|
GenBankmRNA | BY717346.1 | - | 264 | Chr. - - [-] on Build | 94 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | RIKEN cDNA 2810403D21 gene | 2810403D21Rik |
1810022M18Rik, 4632410B05Rik, AU017716 |
-114.6kb | Sense | Chr. X - 108834478 - 108886896 on NCBI Build GRCm38 |
Downstream | high-mobility group nucleosome binding domain 5 | Hmgn5 |
GARP45, NBP-45, Nsbp1 |
2.2kb | Anti-sense | Chr. X - 109004537 - 109013380 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Mus musculus 1700 Both primers and probe map within a single exon |