Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp2c53-ps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_033614.1 | 6 - 7 | 890 | 135 | ||
GenBank mRNA | AY227735.1 | - | 6 - 7 | 890 | 135 |
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream |
cytochrome P450, family 2, subfamily c, polypeptide 66 |
Cyp2c66 | 2010301M18Rik | -66.6kb | Sense | Chr. 19 - 39113898 - 39186756 on NCBI Build GRCm38 |
Downstream |
cytochrome P450, family 2, subfamily c, polypeptide 29 |
Cyp2c29 |
AHOH, AHOHase, AI159681, AI196010, Ah-2, Ahh-1, Cyp2c, P450-2C |
15.4kb | Sense | Chr. 19 - 39287044 - 39330713 on NCBI Build GRCm38 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mus musculus Probe spans exons |