Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008096.2 | NP_032122.1 | 11 - 12 | 1574 | 101 | |
GenBank mRNA | AI876501.1 | - | 2 - 3 | 101 | ||
AK010965.1 | - | 11 - 12 | 1398 | 101 | ||
BC010762.1 | - | 11 - 12 | 1437 | 4234788 | 101 | |
BC051395.1 | - | 11 - 12 | 1441 | 5125043 | 101 | |
M55413.1 | - | 11 - 12 | 1384 | 101 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |